Learn More
Sino Biological FEN1 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 100916T4650
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the human FEN1, and purified by antigen affinity chromatography.
Catalog No. 50-161-2375