Learn More
Sino Biological FOS / c-Fos Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 100918T08100
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human FOS / c-Fos, and purified by antigen affinity chromatography.
Catalog No. 50-161-2382