Learn More
Sino Biological GAS2 / growth arrest-specific 2 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 100922T02200
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human GAS2 / growth arrest-specific 2, and purified by antigen affinity chromatography.
Catalog No. 50-161-2395