Learn More
Sino Biological IGF2BP2/IMP2 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 100932T1050
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human IGF2BP2/IMP2, and purified by antigen affinity chromatography.
Catalog No. 50-161-2420