Learn More
Sino Biological GAPDH Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 10094T16100
Produced in rabbits immunized with purified, recombinant Human GAPDH (rh GAPDH; Catalog#10094-H07E; P04406; Met1-Glu335). GAPDH specific IgG was purified by Human GAPDH affinity chromatography.
Catalog No. 50-161-2451