Learn More
Sino Biological B4GALNT1 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 100942T0850
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human B4GALNT1, and purified by antigen affinity chromatography.
Catalog No. 50-161-2432