Learn More
Sino Biological cubilin / CUBN Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 100948T40200
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human cubilin / CUBN, and purified by antigen affinity chromatography.
Catalog No. 50-161-2446