Learn More
Sino Biological MAP2K3 / MKK3 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 100952T36100
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human MAP2K3 / MKK3, and purified by antigen affinity chromatography.
Catalog No. 50-161-2463