Learn More
Sino Biological MAP2K5 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 100953T10200
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human MAP2K5, and purified by antigen affinity chromatography.
Catalog No. 50-161-2470