Learn More
Sino Biological JNK1 / MAPK8 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 100960T10200
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human JNK1 / MAPK8, and purified by antigen affinity chromatography.
Catalog No. 50-161-2488