Learn More
Sino Biological JNK2/MAPK9 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 100961T1050
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human JNK2/MAPK9, and purified by antigen affinity chromatography.
Catalog No. 50-161-2495