Learn More
Sino Biological MECOM Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 100966T34200
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human MECOM, and purified by antigen affinity chromatography.
Catalog No. 50-161-2500