Learn More
Sino Biological MSH6 / GTBP Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 100973T08100
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the human MSH6, and purified by antigen affinity chromatography.
Catalog No. 50-161-2519