Learn More
Sino Biological Human IL-8 / CXCL8 Protein (aa 23-99) DFS Item
Manufacturer:  Sino Biological 10098HNCH150
The 77 amino acid endothelial-cell derived form (Ala 23-Ser 99) of the mature human IL8 (NP_000575.1) was expressed and purified.
Catalog No. 50-161-3990