Learn More
Sino Biological IL-8/Interleukin-8/CXCL8 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 10098T36200
Produced in rabbits immunized with purified, recombinant Human IL-8/Interleukin-8/CXCL8 (rh IL-8/Interleukin-8/CXCL8; Catalog#10098-HNCH1; NP_000575.1; Ala23-Ser99). IL-8/Interleukin-8/CXCL8 specific IgG was purified by Human IL-8/Interleukin-8/CXCL8 affinity chromatography.
Catalog No. 50-161-2552