Learn More
Sino Biological PAK1 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 100983T32200
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the human PAK1, and purified by antigen affinity chromatography.
Catalog No. 50-161-2532