Learn More
Sino Biological PHF2 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 100989T32200
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human PHF2, and purified by antigen affinity chromatography.
Catalog No. 50-161-2538