Learn More
Sino Biological PS6K / RPS6KB1 Antibody, Mouse MAb DFS Item
Manufacturer:  Sino Biological 10099MM05100
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human PS6K / RPS6KB1 (rh PS6K / RPS6KB1; Catalog#10099-H09B; P23443; Met1-Leu525). The IgG fraction of the cell culture supernatant was purified by Protein A affinity.
Catalog No. 50-161-2578