Learn More
Sino Biological PS6K / RPS6KB1 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 10099T38200
Produced in rabbits immunized with purified, recombinant Human PS6K / RPS6KB1 (rh PS6K / RPS6KB1; Catalog#10099-H09B; P23443; Met1-Leu525). PS6K / RPS6KB1 specific IgG was purified by Human PS6K / RPS6KB1 affinity chromatography.
Catalog No. 50-161-2582