Learn More
Sino Biological PML/RNF71/TRIM19 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 100997T38200
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human PML/RNF71/TRIM19, and purified by antigen affinity chromatography.
Catalog No. 50-161-2573