Learn More
DOPAK INC 100CC CLR RUB/ALUM CAPS 84CS DFS Item
Manufacturer:  DOPAK INC 1900100
Set of 84 pcs. 100 cc clear glass bottle with aluminium cap pp28 and natural rubber septum.
Catalog No. NC0108010