Learn More
Tecan 100mL Disposable Trough
Supplier: Tecan 10613048
Made of natural clear polypropylene and is autoclavable. Bottom of trough has a bevel angle. Dead volume approximately 3. 3 mL. Scaling on trough every 10 mL from 10 - 100 mL. For use in Tecan carrier p/n 10613020. Tecan STERILE purity level-tested free from human DNA, Dnase, Rnase, pyroxenes and endotoxins by an independent lab and guaranteed by a Certificate of Purity. 18 bags of 6 - 108 troughs/sales unit.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.