Learn More
Marathon LS 100μL Universal Pipette Tips, Low Retention, Filter, Sterile, 96 tips/rack, 10 racks/pack.
SDP
Supplier:  Marathon LS MLSPTFS100
Encompass_Preferred
100μL Universal Pipette Tips, Low Retention, Filter, Sterile, 96 tips/rack, 10 racks/pack. Lot traceable and DNase free, RNase free, and non-pyrogenic. Additional info on MLS-PTFS-100: Sterile & Super low retention. Made of clear, high-quality polypropylene. Molded volume graduation markings. Interior support ribs prevent rack from warping. Smooth, hydrophobic surfaces to ensure accuracy. Lids have a stacking feature for stability and a secure locking tab. Packaged in yellow rigid autoclavable racks. Filters reduce cross contamination and prevent damaging aerosols from penetrating into the pipette. MADE IN THE USA. Certifications: ISO13485:2016& ISO9001:2015. These pipette tips are universal fit and are compatible with major pipettes, including Eppendorf and Corning.
Catalog No. NC2609868
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.