Learn More
Athens Research And Technology IncSupplier Diversity Partner 100UG CALPAIN-1 PORCINE ERYTHR DFS Item
Manufacturer:  Athens Research And Technology IncSupplier Diversity Partner 1605030112P100UG
Calpain-1, Porcine Erythrocyte: 100 UG; Frozen in 20 mM imidazole-HCl, 5 mM Beta -mercaptoethanol, 1 mM EDTA, 1mM EGTA, 30% glycerol, pH 6.8; MW 110,000; Activity: > = 1000 units/mg protein. Purified from Porcine Erythrocytes.
Catalog No. 50-490-009