Learn More
Leap PAL PartsSupplier Diversity Partner L-MARK Brand 100l Syringe, FN, GT, Pt. AS, 26s, 2 Pack  DFS Item
Manufacturer:  Leap PAL PartsSupplier Diversity Partner LMK.26207062P
L-MARK Brand 100l, L-MARK Brand Syringe, FN, GT, 26gs, NDL Length 51mm, Pt. Style AS, Plunger Tip PTFE, 2 Pack Made in Germany.
Catalog No. 50-635-321