Learn More
LW Scientific™ ZIP IQ Combo Centrifuge
The Zip-IQ Combo is very small, conserving valuable counter space in the lab, but also sturdy and heavy with suction-cupped feet for stability. Unique smart IQ technology senses which rotor is installed and automatically limits the maximum speed of each rotor for safety.
Specifications
Controller Type Front Display Panel
Display LCD
Electrical Requirements 110-240V
For Use With (Equipment) Laboratory Equipment
Includes Test Tube Rotor, Hematocrit Rotor
View More Specs
Products 2
Catalog Number Mfr. No. Rotor Price Quantity & Availability  
Catalog Number Mfr. No. Rotor Price Quantity & Availability  
22-500-122
View Documents
LW Scientific
ZICCMBO4RT3
2 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
22-500-121
View Documents
LW Scientific
ZICCMBO2RT3
4 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
  • P1 and P2 programmable buttons for time and speed
  • Brushless maintenance-free motor
  • Smooth, whisper-quiet operation
  • Digital time and speed for accuracy
  • Small footprint conserves counter space
Specifications
Specifications
Front Display Panel
110-240V
Test Tube Rotor, Hematocrit Rotor
Zip IQ Combo
No
Yes
Yes
LCD
Laboratory Equipment
Brushless
1
Yes
Combination Centrifuge