Learn More
Sino Biological CD84 / SLAMF5 Antibody, Rabbit PAb DFS Item
Manufacturer:  Sino Biological 10100RP01200
Produced in rabbits immunized with purified, recombinant Human CD84 extracellular domain(rh CD84; Catalog#10100-H08H; Met 1-Gly 225; NP_003865.1). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-161-2618