Learn More
Sino Biological Ppp1r8 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 101001T32200
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human Ppp1r8, and purified by antigen affinity chromatography.
Catalog No. 50-161-2585