Learn More
Sino Biological TLK1 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 101007T3850
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human TLK1, and purified by antigen affinity chromatography.
Catalog No. 50-161-2601