Learn More
Sino Biological Human CD55 / DAF Protein (Fc Tag) DFS Item
Manufacturer:  Sino Biological 10101H02H50
A DNA sequence encoding the human CD55 precursor (NP_000565.1) (Met 1-Ser 353) was expressed with the C-terminal fused Fc region of human IgG1.
Catalog No. 50-161-3998