Learn More
Sino Biological TRAF1 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 101010T32100
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the human TRAF1, and purified by antigen affinity chromatography.
Catalog No. 50-161-2623