Learn More
Sino Biological TRAF6 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 101013T38200
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human TRAF6, and purified by antigen affinity chromatography.
Catalog No. 50-161-2633