Learn More
Sino Biological CD34 Antibody, Rabbit PAb DFS Item
Manufacturer:  Sino Biological 10103RP01400
Produced in rabbits immunized with purified, recombinant Human CD34 (rh CD34; Catalog#10103-H08H; NP_001020280.1; Met1-Thr290). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-161-2734