Learn More
Sino Biological PRKDC Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 101038T1050
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human PRKDC, and purified by antigen affinity chromatography.
Catalog No. 50-161-2726