Learn More
Sino Biological PRMT2 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 101039T10200
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human PRMT2, and purified by antigen affinity chromatography.
Catalog No. 50-161-2728