Learn More
Sino Biological Human IL1RL1 / DER4 Protein (Fc Tag) DFS Item
Manufacturer:  Sino Biological 10105H02H200
A DNA sequence encoding the human IL1RL1 isoform 2 (NP_003847.2) (Met 1-Phe 328) was fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-4006