Learn More
Sino Biological Human IL1RL1 / DER4 Protein DFS Item
Manufacturer:  Sino Biological 10105HCCH50
The secreted isoform of human IL1RL1 (NP_003847.2) (Met 1-Phe 328) with five amino acids (DDDDK) at the C-terminus was expressed and purified.
Catalog No. 50-161-4010