Learn More
Sino Biological IL1RL1 / IL‑1 R4 Antibody, Mouse MAb DFS Item
Manufacturer:  Sino Biological 10105MM12200
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human IL1RL1 / IL‑1 R4 (rh IL1RL1 / IL‑1 R4; Catalog#10105-H08H; NP_003847.2; Met1-Phe328). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-172-0981