Learn More
Sino Biological IL1RL1 Antibody, Rabbit PAb DFS Item
Manufacturer:  Sino Biological 10105RP01200
Produced in rabbits immunized with purified, recombinant Human IL1RL1 / ST2 (rh IL1RL1 / ST2; Catalog#10105-HCCH; aa 1-328; NP_003847.2). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-172-0986