Learn More
Sino Biological IL1RL1 / ST2 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 10105RP03100
Produced in rabbits immunized with purified, recombinant Human IL1RL1 extracellular domain (rhIL1RL1; Catalog#10105-HCCH; aa 1-328; NP_003847.2). IL1RL1 specific IgG was purified by human IL1RL1 affinity chromatography.
Catalog No. 50-172-0988