Learn More
Sino Biological Carbonic Anhydrase IX/CA9 Antibody, Mouse MAb DFS Item
Manufacturer:  Sino Biological 10107MM02200
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Carbonic Anhydrase IX/CA9 (rh Carbonic Anhydrase IX/CA9; Catalog#10107-H08H; NP_001207.2; Met1-Asp414). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-172-1016