Learn More
Sino Biological Carbonic Anhydrase IX / CA9 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 10107T48200
Produced in rabbits immunized with purified, recombinant Human Carbonic Anhydrase IX / CA9 (rh Carbonic Anhydrase IX / CA9; Catalog#10107-H08H; NP_001207.2; Met1-Asp414). Carbonic Anhydrase IX / CA9 specific IgG was purified by Human Carbonic Anhydrase IX / CA9 affinity chromatography.
Catalog No. 50-172-1039