Learn More
Sino Biological ACE2 / Angiotensin-Converting Enzyme 2 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 10108T56200
Produced in rabbits immunized with purified, recombinant Human ACE2 / Angiotensin-Converting Enzyme 2 (rh ACE2 / Angiotensin-Converting Enzyme 2; NP_068576.1; 1M-740S). ACE2 / Angiotensin-Converting Enzyme 2 specific IgG was purified by Human ACE2 / Angiotensin-Converting Enzyme 2 affinity chromatography.
Catalog No. 50-172-1066