Learn More
Sino Biological STAT4 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 101081T34100
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human STAT4, and purified by antigen affinity chromatography.
Catalog No. 50-172-1044