Learn More
Sino Biological STUB1 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 101082T08200
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human STUB1, and purified by antigen affinity chromatography.
Catalog No. 50-172-1048