Learn More
Sino Biological SUMO2 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 101083T38200
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human SUMO2, and purified by antigen affinity chromatography.
Catalog No. 50-172-1051