Learn More
Sino Biological Human CD155 / PVR Protein (His Tag), Biotinylated DFS Item
Manufacturer:  Sino Biological 10109H08HB20
A DNA sequence encoding the PVR (NP_006496.3) (Met1-Asn343) was expressed with a C-terminal polyhistidine tag. The purified protein was biotinylated in vitro.
Catalog No. 50-161-4027