Learn More
Sino Biological CD155 / PVR Antibody, Mouse MAb DFS Item
Manufacturer:  Sino Biological 10109MM11100
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD155 extracellular domain (rh CD155; Catalog#10109-H08H; Met 1-Asn 343; NP_006496.2). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-172-1080