Learn More
Sino Biological Human IL1R2 / CD121b Protein (Fc Tag) DFS Item
Manufacturer:  Sino Biological 10111H02H50
A DNA sequence encoding the extracellular domain (Met 1-Glu 343) of human IL1 receptor type I I (IL1RII) (NP_004624.1) was expressed with C-terminal fused Fc region of human IgG1.
Catalog No. 50-161-4031