Learn More
Sino Biological IL-1R2 / CD121b Antibody, Mouse MAb DFS Item
Manufacturer:  Sino Biological 10111MM0250
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human IL-1R2 / CD121b (rh IL-1R2 / CD121b; Catalog#10111-H08H; NP_004624.1; Met1-Glu343). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-172-1132